Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128170.2 | NP_001121642.1 | 15 | 2473 | ||
NM_001128171.2 | NP_001121643.1 | 15 | 2598 | |||
NM_001276279.1 | NP_001263208.1 | 13 | 2049 | |||
NM_173369.3 | NP_775545.1 | 14 | 2464 | |||
GenBank mRNA | AK122389.1 | - | 15 | 2579 | ||
AK133479.1 | - | 14 | 2428 | |||
AK134926.1 | - | 2 | 156 | |||
AK147204.1 | - | 15 | 2442 | |||
AK147426.1 | - | 15 | 2437 | |||
AK167424.1 | - | 13 | 2013 | |||
BC024596.1 | - | 2 | 142 | |||
BC028885.1 | - | 7 | 2119 | |||
BC042438.1 | - | 14 | 2417 | 3983771 | ||
BC049879.1 | - | 5 | 326 |