Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128170.2 | NP_001121642.1 | 14 | 2430 | ||
NM_001128171.2 | NP_001121643.1 | 14 | 2555 | |||
NM_001276279.1 | NP_001263208.1 | 12 | 2006 | |||
NM_173369.3 | NP_775545.1 | 13 | 2421 | |||
GenBank mRNA | AK122389.1 | - | 14 | 2536 | ||
AK133479.1 | - | 13 | 2385 | |||
AK134926.1 | - | 1 | 113 | |||
AK147204.1 | - | 14 | 2399 | |||
AK147426.1 | - | 14 | 2394 | |||
AK167424.1 | - | 12 | 1970 | |||
BC024596.1 | - | 1 | 99 | |||
BC028885.1 | - | 6 | 2076 | |||
BC042438.1 | - | 13 | 2374 | 3983771 | ||
BC049879.1 | - | 4 | 283 |