Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028841.3 | NP_083117.2 | 5 | 679 | ||
GenBank mRNA | AK008761.1 | - | 5 | 624 | ||
AK166272.1 | - | 5 | 682 | |||
AK171691.1 | - | 5 | 628 | |||
AK219107.1 | - | 2 | 117 | |||
BC010346.1 | - | 5 | 624 | 2811935 |