Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028846.5 | NP_083122.1 | 3 | 304 | ||
GenBank mRNA | AF449715.1 | - | 3 | 255 | ||
AK029890.1 | - | 201 | ||||
AK042612.1 | - | 3 | 221 | |||
AK047242.1 | - | 3 | 225 | |||
AK054279.1 | - | 3 | 247 | |||
AK087412.1 | - | 3 | 215 | |||
AK163663.1 | - | 3 | 214 | |||
AK173083.1 | - | 3 | 206 | |||
BC079674.1 | - | 193 | 30531994 | |||
BY251374.1 | - | 3 | 305 |