Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297418.1 | NP_001284347.1 | 7 | 927 | ||
NM_005968.4 | NP_005959.2 | 8 | 889 | |||
NM_031203.3 | NP_112480.2 | 9 | 772 | |||
XM_005272478.2 | XP_005272535.1 | 8 | 1233 | |||
XM_005272479.2 | XP_005272536.1 | 8 | 1233 | |||
XM_005272480.2 | XP_005272537.1 | 9 | 1116 | |||
XM_005272481.1 | XP_005272538.1 | 7 | 835 | |||
XM_005272483.2 | XP_005272540.1 | 8 | 700 | |||
XM_017026823.1 | XP_016882312.1 | 8 | 1233 | |||
XM_017026824.1 | XP_016882313.1 | 9 | 1116 | |||
XM_017026825.1 | XP_016882314.1 | 9 | 1116 | |||
XM_017026826.1 | XP_016882315.1 | 7 | 780 | |||
XM_017026827.1 | XP_016882316.1 | 7 | 835 | |||
XM_017026828.1 | XP_016882317.1 | 8 | 667 | |||
XM_017026829.1 | XP_016882318.1 | 8 | 718 | |||
XM_017026830.1 | XP_016882319.1 | 8 | 718 | |||
XM_017026831.1 | XP_016882320.1 | 8 | 687 | |||
XM_017026832.1 | XP_016882321.1 | 8 | 687 | |||
GenBank mRNA | AB209733.1 | - | 7 | 755 | ||
AF061832.1 | - | 9 | 922 | |||
AK309659.1 | - | 838 | ||||
BC000138.2 | - | 8 | 850 | 2900532 | ||
BC019580.1 | - | 8 | 818 | 4938259 | ||
L03532.1 | - | 8 | 817 | |||
L32611.1 | - | 9 | 701 | |||
X72018.1 | - | 4 | 455 |