Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004536.2 | NP_004527.2 | 12 | 2507 | ||
NM_022892.1 | NP_075043.1 | 10 | 1836 | |||
XM_011543413.2 | XP_011541715.1 | 2 | 645 | |||
XM_017009490.1 | XP_016864979.1 | 2 | 1192 | |||
GenBank mRNA | AB048534.1 | - | 10 | 1836 | ||
AB209123.1 | - | 1 | 1893 | |||
AK124511.1 | - | 1 | 1188 | |||
BC136273.1 | - | 10 | 1844 | 9020260 | ||
BC143761.1 | - | 10 | 1844 | 9052275 | ||
DQ786305.1 | - | 1 | 153 | |||
U19251.1 | - | 12 | 2080 |