Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028870.3 | NP_083146.1 | 2 | 222 | ||
GenBank mRNA | AK029382.1 | - | 2 | 223 | ||
AK045989.1 | - | 2 | 243 | |||
AK146816.1 | - | 295 | ||||
AK159480.1 | - | 2 | 272 | |||
AK167531.1 | - | 295 | ||||
BC070404.1 | - | 2 | 224 | 5699653 |