Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277121.1 | NP_001264050.1 | 5 | 568 | ||
NM_001277122.1 | NP_001264051.1 | 4 | 407 | |||
NM_001277123.1 | NP_001264052.1 | 4 | 402 | |||
NM_028871.2 | NP_083147.1 | 5 | 571 | |||
GenBank mRNA | AF441128.1 | - | 4 | 411 | ||
AK011307.1 | - | 4 | 320 | |||
AK133840.1 | - | 4 | 328 | |||
AK141145.1 | - | 5 | 516 | |||
AK144682.1 | - | 5 | 521 | |||
AK144992.1 | - | 4 | 325 | |||
AK145849.1 | - | 4 | 346 | |||
AK147043.1 | - | 5 | 537 | |||
AK152039.1 | - | 5 | 543 | |||
AK163805.1 | - | 1 | 872 | |||
AK181037.1 | - | 3 | 264 | |||
BC004679.1 | - | 4 | 319 | |||
BC026850.1 | - | 5 | 504 | |||
BC038051.1 | - | 5 | 548 | 3500728 |