Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145824.4 | NP_665823.2 | 13 | 1799 | ||
GenBank mRNA | AK087165.1 | - | 3 | 216 | ||
AK173177.1 | - | 13 | 1612 | |||
AY337314.1 | - | 13 | 1800 | |||
BC024698.1 | - | 13 | 1644 | 3667000 | ||
EU281316.1 | - | 13 | 1660 |