Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028881.2 | NP_083157.1 | 11 | 1483 | ||
GenBank mRNA | AA611164.1 | - | 1 | 203 | ||
AK014553.1 | - | 6 | 1854 | |||
AK156192.1 | - | 11 | 1462 | |||
AK169944.1 | - | 7 | 1132 | |||
BC023091.1 | - | 7 | 1073 | 4023064 | ||
BC023831.1 | - | 11 | 1478 | 5345301 | ||
BC032183.1 | - | 11 | 1488 | 5310152 | ||
BC033300.1 | - | 11 | 1478 | 4166411 |