Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242363.2 | NP_001229292.1 | 10 | 1920 | ||
NM_001242364.2 | NP_001229293.1 | 9 | 1871 | |||
NM_001242365.2 | NP_001229294.1 | 9 | 1871 | |||
NM_199009.3 | NP_950174.1 | 9 | 1829 | |||
GenBank mRNA | AK085350.1 | - | 9 | 1824 | ||
AK122552.1 | - | 9 | 1757 | |||
AK154173.1 | - | 10 | 1864 | |||
AK155273.1 | - | 10 | 1865 | |||
AK159716.1 | - | 9 | 1826 | |||
AK169996.1 | - | 10 | 1902 | |||
BC018464.1 | - | 9 | 1788 | |||
BC021532.1 | - | 7 | 1198 | |||
BC043717.1 | - | 8 | 1345 | |||
BC063093.1 | - | 9 | 1786 | 6332038 |