Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028894.1 | NP_083170.1 | 1 | 980 | ||
GenBank mRNA | AK016522.1 | - | 1 | 980 | ||
AK030866.1 | - | 1 | 790 | |||
AK147751.1 | - | 1 | 984 | |||
AK167341.1 | - | 1 | 1038 | |||
AK177253.1 | - | 1 | 44 | |||
AK189559.1 | - | 1 | 44 | |||
BC120688.1 | - | 1 | 882 | 40129611 | ||
BC120690.1 | - | 1 | 882 | 40129613 |