Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028894.1 | NP_083170.1 | 10 | 2220 | ||
GenBank mRNA | AK016522.1 | - | 10 | 2220 | ||
AK167341.1 | - | 10 | 2279 | |||
AK185175.1 | - | 3 | 315 | |||
BC120688.1 | - | 10 | 2122 | 40129611 | ||
BC120690.1 | - | 10 | 2122 | 40129613 |