Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mei1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028897.3 | NP_083173.2 | 30 | 3726 | ||
GenBank mRNA | AK016514.1 | - | 24 | 2931 | ||
AY270177.1 | - | 23 | 3472 | |||
BC137749.1 | - | 23 | 2759 | 8860768 | ||
BC144888.1 | - | 22 | 2661 | |||
BY641487.1 | - | 2 | 142 |