Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB082952.1 | - | 4 | 443 | ||
AK016530.1 | - | 4 | 590 | |||
AK029341.1 | - | 4 | 1239 | |||
AK087217.1 | - | 4 | 1259 | |||
AK144082.1 | - | 4 | 1238 | |||
AK161011.1 | - | 4 | 1349 | |||
AK172276.1 | - | 4 | 1236 | |||
BC060140.1 | - | 4 | 555 | |||
BC118971.1 | - | 4 | 438 | |||
BC148309.2 | - | 4 | 443 |