Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204229.1 | NP_001191158.1 | 23 | 3036 | ||
NM_177562.5 | NP_808230.2 | 24 | 3042 | |||
GenBank mRNA | AK016529.1 | - | 15 | 1855 | ||
AK122255.1 | - | 24 | 2994 | |||
AK156927.1 | - | 23 | 2967 | |||
AK171134.1 | - | 22 | 2894 | |||
BC138907.1 | - | 24 | 2945 | 8861929 | ||
BC138909.1 | - | 24 | 2945 | 8861931 |