Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204229.1 | NP_001191158.1 | 5 | 807 | ||
NM_177562.5 | NP_808230.2 | 5 | 807 | |||
GenBank mRNA | AK089327.1 | - | 5 | 737 | ||
AK122255.1 | - | 5 | 759 | |||
AK155314.1 | - | 5 | 749 | |||
AK156927.1 | - | 5 | 738 | |||
AK170754.1 | - | 5 | 738 | |||
AK171134.1 | - | 5 | 738 | |||
BC138907.1 | - | 5 | 710 | 8861929 | ||
BC138909.1 | - | 5 | 710 | 8861931 |