Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165983.1 | NP_001159455.1 | 13 | 1379 | ||
NM_001165984.1 | NP_001159456.1 | 13 | 1339 | |||
NM_001165985.1 | NP_001159457.1 | 13 | 1397 | |||
NM_001165986.1 | NP_001159458.1 | 13 | 1372 | |||
NM_001165987.1 | NP_001159459.1 | 12 | 1304 | |||
NM_001165988.1 | NP_001159460.1 | 12 | 1279 | |||
NM_028475.2 | NP_082751.1 | 13 | 1364 | |||
NM_153489.2 | NP_705693.2 | 12 | 1304 | |||
GenBank mRNA | AF276967.1 | - | 12 | 1239 | ||
AK016538.1 | - | 6 | 631 | |||
AK030218.1 | - | 13 | 1305 | |||
AK031934.1 | - | 12 | 1284 | |||
AK048286.1 | - | 13 | 1380 | |||
AK051093.1 | - | 3 | 1182 | |||
AK079895.1 | - | 13 | 1333 | |||
AK082804.1 | - | 4 | 483 | |||
AK088702.1 | - | 13 | 1395 | |||
BC023906.1 | - | 12 | 1295 | 5321969 | ||
BC029075.1 | - | 13 | 1354 | 4527901 | ||
BC050910.1 | - | 12 | 1286 | 6417527 |