Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145826.1 | NP_001139298.1 | 17 | 3564 | ||
NM_153406.3 | NP_700455.3 | 14 | 3335 | |||
GenBank mRNA | AB093236.1 | - | 17 | 3564 | ||
AK015541.1 | - | 3 | 300 | |||
AK016555.1 | - | 3 | 301 | |||
AY884297.1 | - | 14 | 3336 | |||
BC037464.1 | - | 11 | 2422 | 4194868 | ||
BC072595.1 | - | 13 | 3174 | 6408832 |