Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028932.4 | NP_083208.1 | 2 | 361 | ||
GenBank mRNA | AK016628.1 | - | 1042 | |||
AK036661.1 | - | 2 | 333 | |||
AK044181.1 | - | 2 | 362 | |||
AK044737.1 | - | 2 | 298 | |||
BC079658.1 | - | 2 | 338 | 6853920 |