Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933402E13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199996.1 | NP_001186925.1 | 4 | 983 | ||
NM_001199997.1 | NP_001186926.1 | 4 | 944 | |||
GenBank mRNA | AK016614.1 | - | 4 | 986 | ||
BC132156.1 | - | 4 | 979 | 40130433 |