Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029912.2 | NP_001025083.1 | 5, 6 | 1503 | ||
GenBank mRNA | AK122519.1 | - | 5, 6 | 1503 |