Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933427G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028955.1 | NP_083231.1 | 5 | 1393 | ||
GenBank mRNA | AK016951.1 | - | 5 | 1393 |