Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033654.2 | NP_001028826.1 | 17 | 1582 | ||
NM_028304.2 | NP_082580.1 | 17 | 1995 | |||
NM_028956.4 | NP_083232.1 | 17 | 1592 | |||
GenBank mRNA | AK017063.1 | - | 17 | 1569 | ||
AK028368.1 | - | 17 | 1579 | |||
AK029866.1 | - | 17 | 1583 | |||
AK088562.1 | - | 17 | 1583 | |||
AK145055.1 | - | 17 | 1941 | |||
BC025555.1 | - | 17 | 1813 | 5321811 |