Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163389.1 | NP_001156861.1 | 6 | 488 | ||
NM_028965.4 | NP_083241.1 | 7 | 548 | |||
GenBank mRNA | AK017118.1 | - | 2 | 154 | ||
AK042409.1 | - | 6 | 464 | |||
AK076332.1 | - | 7 | 524 | |||
BC014719.1 | - | 6 | 465 | 4236099 |