Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204084.2 | NP_001191013.1 | 4 | 1688 | ||
NM_001204085.1 | NP_001191014.1 | 4 | 1606 | |||
NM_001204086.2 | NP_001191015.1 | 4 | 1715 | |||
NM_001278164.1 | NP_001265093.1 | 4 | 1597 | |||
NM_001278165.1 | NP_001265094.1 | 4 | 1664 | |||
NM_001278166.1 | NP_001265095.1 | 4 | 1508 | |||
NM_005380.7 | NP_005371.2 | 4 | 1721 | |||
NM_182744.3 | NP_877421.2 | 4 | 1616 | |||
GenBank mRNA | BC012037.1 | - | 4 | 1520 | 3874204 | |
D28124.1 | - | 4 | 1489 |
Gene Symbol: MINOS1-NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204088.1 | NP_001191017.1 | 5 | 1765 | ||
NM_001204089.1 | NP_001191018.1 | 4 | 1580 | |||
GenBank mRNA | AK289456.1 | - | 4 | 1581 |