Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159288.1 | NP_001152760.1 | 24 | 3406 | ||
NM_198162.1 | NP_937805.1 | 23 | 2736 | |||
GenBank mRNA | AK018377.1 | - | 4 | 480 | ||
AK173042.1 | - | 24 | 3406 | |||
BC059243.1 | - | 23 | 2736 | 6417551 | ||
BC063082.1 | - | 12 | 1673 |