Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 8430419L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028982.4 | NP_083258.2 | 12 | 1911 | ||
GenBank mRNA | AK018428.1 | - | 12 | 1855 | ||
AK039401.1 | - | 12 | 1847 | |||
AK155128.1 | - | 12 | 1856 | |||
AK220395.1 | - | 11 | 1561 | |||
BC003918.1 | - | 4 | 510 | |||
BC038002.1 | - | 11 | 1572 | |||
BC052213.1 | - | 1 | 1699 | |||
BC079676.1 | - | 12 | 1838 | |||
BC137674.1 | - | 12 | 1890 | 8860693 | ||
BC137676.1 | - | 12 | 1890 | 8860695 |