Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028994.2 | NP_083270.1 | 10 | 1897 | ||
GenBank mRNA | AK007429.1 | - | 7 | 1089 | ||
AK030501.1 | - | 10 | 1898 | |||
AK031612.1 | - | 10 | 1742 | |||
AK034927.1 | - | 10 | 1776 | |||
AK142561.1 | - | 4 | 649 | |||
AK147972.1 | - | 10 | 1751 | |||
AK165094.1 | - | 10 | 1775 | |||
AK170242.1 | - | 10 | 1751 | |||
AK199613.1 | - | 1 | 55 | |||
BC010318.1 | - | 10 | 1760 | 3596227 | ||
BC011126.1 | - | 9 | 2059 | |||
BC023437.1 | - | 8 | 1365 | |||
BC028326.1 | - | 5 | 698 |