Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028994.2 | NP_083270.1 | 9, 10 | 1696 | ||
GenBank mRNA | AK007429.1 | - | 6, 7 | 888 | ||
AK030501.1 | - | 9, 10 | 1697 | |||
AK031612.1 | - | 9, 10 | 1541 | |||
AK034927.1 | - | 9, 10 | 1575 | |||
AK142561.1 | - | 3, 4 | 448 | |||
AK147972.1 | - | 9, 10 | 1550 | |||
AK165094.1 | - | 9, 10 | 1574 | |||
AK170242.1 | - | 9, 10 | 1550 | |||
AK218547.1 | - | 2, 3 | 235 | |||
BC010318.1 | - | 9, 10 | 1559 | 3596227 | ||
BC011126.1 | - | 8, 9 | 1858 | |||
BC023437.1 | - | 7, 8 | 1164 | |||
BC028326.1 | - | 4, 5 | 497 |