Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029001.5 | NP_083277.3 | 3 | 373 | ||
GenBank mRNA | AK018616.1 | - | 3 | 348 | ||
AK037162.1 | - | 3 | 345 | |||
AK049118.1 | - | 3 | 373 | |||
AK137668.1 | - | 3 | 358 | |||
BC005602.1 | - | 3 | 373 | 3483017 |