Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000615.6 | NP_000606.3 | 10 | 1678 | ||
NM_001076682.3 | NP_001070150.1 | 10 | 1678 | |||
NM_001242607.1 | NP_001229536.1 | 12 | 1786 | |||
NM_001242608.1 | NP_001229537.1 | 10 | 1678 | |||
NM_181351.4 | NP_851996.2 | 11 | 1708 | |||
GenBank mRNA | AB209443.1 | - | 10 | 1557 | ||
AK057509.1 | - | 10 | 1781 | |||
AK292453.1 | - | 11 | 1343 | |||
AK314589.1 | - | 12 | 1758 | |||
BC014205.2 | - | 5 | 572 | |||
BC047244.1 | - | 11 | 1520 | 5590188 | ||
S71824.1 | - | 10 | 1513 | |||
U63041.1 | - | 10 | 1349 | |||
X16841.1 | - | 10 | 1457 |