Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyroxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029011.2 | NP_083287.2 | 13 | 1635 | ||
GenBank mRNA | AK154222.1 | - | 13 | 1397 | ||
AK160239.1 | - | 13 | 1352 | |||
CX218741.1 | - | 6 | 581 |