Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278671.1 | NP_001265600.1 | 2 | 613 | ||
GenBank mRNA | AK014702.1 | - | 2 | 610 | ||
AK016973.1 | - | 2 | 613 | |||
BC139158.1 | - | 1 | 558 | 8862180 | ||
BC139160.1 | - | 1 | 558 | 8862182 |