Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175210.3 | NP_780419.2 | 1 | 383 | ||
GenBank mRNA | AK014713.1 | - | 1 | 371 | ||
AK028781.1 | - | 1 | 384 | |||
AK029031.1 | - | 1 | 384 | |||
BC158053.1 | - | 1 | 196 | 9088162 | ||
BC158063.1 | - | 1 | 196 | 9007362 |