Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173370.3 | NP_775546.2 | 7 | 902 | ||
GenBank mRNA | AF533367.2 | - | 7 | 811 | ||
BC026505.1 | - | 2 | 68 | |||
BC055292.1 | - | 7 | 902 | 6491596 |