Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029017.2 | NP_083293.1 | 6 | 582 | ||
GenBank mRNA | AA174956.1 | - | 1 | 20 | ||
BC029173.1 | - | 562 | ||||
BC038065.1 | - | 6 | 553 | |||
BC087953.1 | - | 6 | 576 | 30119240 | ||
CX122942.1 | - | 6 | 561 |