Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029020.2 | NP_083296.2 | 8, 9 | 1440 | ||
GenBank mRNA | AK014319.1 | - | 8, 9 | 1171 | ||
AK014688.1 | - | 8, 9 | 1193 | |||
AK030624.1 | - | 8, 9 | 1167 | |||
AK038760.1 | - | 8, 9 | 1167 | |||
AK049152.1 | - | 8, 9 | 1184 | |||
AK076315.1 | - | 8, 9 | 1191 | |||
AK166704.1 | - | 8, 9 | 1174 | |||
AK188519.1 | - | 3, 4 | 246 | |||
AK212771.1 | - | 2, 3 | 122 | |||
BC015301.1 | - | 8, 9 | 1440 | 4237894 |