Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc74b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145435.1 | NP_001138907.1 | 3, 4 | 442 | ||
NM_029053.3 | NP_083329.2 | 3, 4 | 442 | |||
GenBank mRNA | AK039092.1 | - | 3, 4 | 420 | ||
AK132588.1 | - | 3, 4 | 440 | |||
AK143245.1 | - | 3, 4 | 415 | |||
AK161226.1 | - | 3, 4 | 440 | |||
BB634732.1 | - | 3, 4 | 443 | |||
BC115678.1 | - | 3, 4 | 419 | 40104166 | ||
BC115679.1 | - | 3, 4 | 419 | 40104167 |