Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127191.2 | NP_001120663.1 | 2 | 578 | ||
NM_029068.4 | NP_083344.3 | 2 | 578 | |||
GenBank mRNA | AK015872.1 | - | 2 | 539 | ||
AK032055.1 | - | 2 | 535 | |||
AK043975.1 | - | 2 | 548 | |||
AK166422.1 | - | 2 | 552 | |||
BC019424.1 | - | 1 | 343 | 5052234 | ||
BC070431.1 | - | 523 |