Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029084.3 | NP_083360.2 | 3 | 761 | ||
GenBank mRNA | AK017911.1 | - | 3 | 664 | ||
AK142633.1 | - | 3 | 762 | |||
BC024587.1 | - | 3 | 681 | 5066283 |