Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024263.4 | NP_077225.4 | 8, 9 | 1382 | ||
GenBank mRNA | AB040488.1 | - | 8, 9 | 1302 | ||
AB052097.1 | - | 8, 9 | 1260 | |||
AK004732.1 | - | 8, 9 | 1265 | |||
BC002140.1 | - | 3, 4 | 379 | |||
BC026438.1 | - | 8, 9 | 1304 | 4239626 | ||
EU849596.1 | - | 8, 9 | 1220 | |||
EU849597.1 | - | 8, 9 | 1211 |