Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290689.1 | NP_001277618.1 | 3, 4 | 423 | ||
NM_029090.3 | NP_083366.1 | 4, 5 | 539 | |||
GenBank mRNA | AK004750.1 | - | 4, 5 | 432 | ||
AK088736.1 | - | 4, 5 | 457 | |||
AK202435.1 | - | 3, 4 | 357 | |||
BC004837.1 | - | 4, 5 | 475 | 2654387 | ||
BY255312.1 | - | 4, 5 | 457 | |||
CF540715.1 | - | 1, 2 | 84 |