Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029091.2 | NP_083367.1 | 12 | 1464 | ||
GenBank mRNA | AK004774.1 | - | 12 | 1465 | ||
AK170793.1 | - | 12 | 1448 | |||
AK182867.1 | - | 2 | 83 |