Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029094.3 | NP_083370.2 | 10 | 1715 | ||
GenBank mRNA | AK081350.1 | - | 10 | 1910 | ||
AK090116.1 | - | 10 | 1592 | |||
AK154106.1 | - | 10 | 1700 | |||
BC127073.1 | - | 9 | 1364 | 40110218 |
Gene Symbol: Gm12992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047558.1 | - | 4 | 792 |