Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930402F06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080709.2 | NP_001074178.1 | 2 | 339 | ||
GenBank mRNA | AK015047.1 | - | 2 | 340 |