Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029116.2 | NP_083392.1 | 2 | 2429 | ||
GenBank mRNA | AK046634.1 | - | 1 | 1501 | ||
AK050286.1 | - | 1 | 1285 | |||
AK080001.1 | - | 1 | 1448 | |||
AK129197.1 | - | 1 | 1392 | |||
BC080858.1 | - | 1 | 1400 | 6852896 |