Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133758.3 | NP_598519.2 | 20 | 2647 | ||
NM_177249.3 | NP_796223.2 | 21 | 2707 | |||
GenBank mRNA | AK030909.1 | - | 5 | 598 | ||
AK050316.1 | - | 1 | 2133 | |||
AK131138.1 | - | 11 | 1324 | |||
BC024117.1 | - | 4 | 473 | |||
BC108425.1 | - | 21 | 2648 | 30062566 |