Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033172.3 | NP_001028344.2 | 4, 5 | 1673 | ||
NM_001164367.1 | NP_001157839.1 | 4, 5 | 1101 | |||
GenBank mRNA | AK015534.1 | - | 4, 5 | 1101 | ||
AK154916.1 | - | 4, 5 | 1673 | |||
BC089010.1 | - | 3, 4 | 932 | |||
BC139380.1 | - | 4, 5 | 1436 | 8862402 | ||
BC139419.1 | - | 4, 5 | 1436 | 8862441 |