Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162863.1 | NP_001156335.1 | 2 | 607 | ||
NM_029145.3 | NP_083421.2 | 2 | 716 | |||
GenBank mRNA | AK015573.1 | - | 2 | 608 | ||
AY226990.1 | - | 2 | 716 | |||
BC141116.1 | - | 2 | 709 | 9055728 |